Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Creatine Kinase BB Mouse anti-Human, Alexa Fluor 700, Clone: 2ba6, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259601AF700
Description
Creatine Kinase BB Monoclonal specifically detects Creatine Kinase BB in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Creatine Kinase BB | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CKB | |
Human Creatine Kinase BB protein (Uniprot: P12277) | |
100 μL | |
Primary | |
The specificity of this monoclonal antibody to its intended target was validated by HuProtTM Array, containing more than 19,000, full-length human proteins. The specificity of this monoclonal antibody to its intended target was validated by HuProt Array containing more than 19,000 full-length, correctly-folded human proteins. Creatine kinases (CK) are a large family of isoenzymes that regulate levels of ATP in subcellular compartments, where they provide ATP at sites of fluctuating energy demand by the transfer of phosphates between creatine and adenine nucleotides. CKs provide the energy of phosphate hydrolysis necessary to drive the normal function of many cellular systems. In cells, the cytosolic CK enzymes consist of two subunits, which can be either B (brain type) or M (muscle type). There are three different isoenzymes: CKMM, CKBB and CKMB. This monoclonal antibody recognizes the CKBB isoenzyme and does not react with the B subunit in CKMB. It shows minimal reactivity with other human serum proteins. | |
Store at 4C in the dark. | |
IgG1 κ |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
2ba6 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
Creatine Kinase Isoenzyme | |
Mouse | |
Protein A or G purified | |
RUO | |
1152 | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)