Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Creatine Kinase BB Mouse anti-Human, Clone: CPTC-CKB-2, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30754220UG
Description
Creatine Kinase BB Monoclonal specifically detects Creatine Kinase BB in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.Specifications
Creatine Kinase BB | |
Monoclonal | |
0.2 mg/ml | |
Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml | |
Creatine Kinase Isoenzyme | |
Recombinant human full-length Creatine Kinase BB protein | |
20 μg | |
Cancer | |
1152 | |
Store at 4°C. | |
IgG2b κ |
Western Blot, Immunohistochemistry (Paraffin) | |
CPTC-CKB-2 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction