Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Creatine Kinase BB Mouse anti-Human, PE/Atto594, Clone: 2ba6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP259601PEATT594

Catalog No. N259601P594


Add to cart

Description

Description

Creatine Kinase BB Monoclonal antibody specifically detects Creatine Kinase BB in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Creatine Kinase BB
2ba6
Flow Cytometry
Conjugated, Purified
Mouse
IgG1 Kappa
0.1 mL
Primary
1152.0
Human
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
CKB
Human CKBB protein
Protein A or G purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
The specificity of this monoclonal antibody to human Creatine Kinase BB was validated by HuProtTM Array, containing more than 19,000, full-length human proteins. The specificity of this monoclonal antibody to human Creatine Kinase BB was validated by HuProt Array containing more than 19,000 full-length, correctly-folded human proteins. Creatine kinases (CK) are a large family of isoenzymes that regulate levels of ATP in subcellular compartments, where they provide ATP at sites of fluctuating energy demand by the transfer of phosphates between creatine and adenine nucleotides. CKs provide the energy of phosphate hydrolysis necessary to drive the normal function of many cellular systems. In cells, the "cytosolic" CK enzymes consist of two subunits, which can be either B (brain type) or M (muscle type). There are three different isoenzymes: CKMM, CKBB and CKMB. This MAb recognizes the CKBB isoenzyme and does not react with the B subunit in CKMB. It shows minimal reactivity with other human serum proteins.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit