Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Creatine Kinase BB, Mouse anti-Human, PE, Clone: CKB/1268, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP275763PE
Description
Creatine Kinase BB Monoclonal antibody specifically detects Creatine Kinase BB in Human samples. It is validated for Flow Cytometry.Specifications
Creatine Kinase BB | |
CKB/1268 | |
Flow Cytometry | |
PBS with 0.05% Sodium Azide | |
CKB | |
Recombinant full-length human Creatine Kinase B protein | |
Monoclonal | |
Protein A or G purified | |
RUO | |
Store at 4°C in the dark. | |
Creatine kinases (CK) are a large family of isoenzymes that regulate levels of ATP in subcellular compartments, where they provide ATP at sites of fluctuating energy demand by the transfer of phosphates between creatine and adenine nucleotides. CKs provide the energy of phosphate hydrolysis necessary to drive the normal function of many cellular systems. In cells, the cytosolic CK enzymes consist of two subunits, which can be either B (brain type) or M (muscle type). There are three different isoenzymes: CKMM, CKBB and CKMB. This MAb recognizes the CKBB (Creatine Kinase B) isoenzyme and does not react with the B subunit in CKMB. It shows minimal reactivity with other human serum proteins |
Flow Cytometry | |
PE | |
Purified,Conjugated | |
Creatine Kinase Isoenzyme | |
Mouse | |
IgG1 Kappa | |
Primary | |
0.1 mL | |
Cancer | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title