Learn More
Invitrogen™ CKMT1B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596224
Description
Immunogen sequence: MAGPFSRLLS ARPGLRLLAL AGAGSLAAGF LLRPEPVRAA SERRRLYPPS AEYPDLRKHN NCMASHLTPA VYARLCDKTT PTGWT; Positive Samples: Mouse brain, Mouse kidney, Rat brain; Cellular Location: Intermembrane side, Mitochondrion inner membrane, Peripheral membrane protein.
Mitochondrial creatine (MtCK) kinase is responsible for the transfer of high energy phosphate from mitochondria to the cytosolic carrier, creatine. It belongs to the creatine kinase isoenzyme family. It exists as two isoenzymes, sarcomeric MtCK and ubiquitous MtCK, encoded by separate genes. Mitochondrial creatine kinase occurs in two different oligomeric forms: dimers and octamers, in contrast to the exclusively dimeric cytosolic creatine kinase isoenzymes. Many malignant cancers with poor prognosis have shown overexpression of ubiquitous mitochondrial creatine kinase; this may be related to high energy turnover and failure to eliminate cancer cells via apoptosis. Ubiquitous mitochondrial creatine kinase has 80% homology with the coding exons of sarcomeric mitochondrial creatine kinase. Two genes located near each other on chromosome 15 have been identified which encode identical mitochondrial creatine kinase proteins.
Specifications
CKMT1B | |
Polyclonal | |
Unconjugated | |
CKMT1B | |
acidic-type mitochondrial creatine kinase; CKMT; CKMT1; CKMT1A; CKMT1B; Creatine kinase U-type, mitochondrial; creatine kinase, mitochondrial 1 (ubiquitous); creatine kinase, mitochondrial 1B; mia-CK; ubiquitous mitochondrial creatine kinase; UMTCK; U-MtCK | |
Rabbit | |
Affinity chromatography | |
RUO | |
1159 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry | |
0.55 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
P12532 | |
CKMT1B | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-85 of human CKMT1B (NP_0662701). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.