Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Creatine Phosphate Disodium Salt Tetrahydrate, ≥98%, Affymetrix/USB™

Click to view available options
Quantity:
5g
25g
Packaging:
Plastic bottle
71519-72-7
C4H16N3Na2O9P
327.14
MFCD00150192
HUWYWJSJJDCZRQ-UHFFFAOYSA-L
creatine phosphate sodium salt,Sodium creatine phosphate dibasic tetrahydrate
131887655
disodium tetrahydrate ({amino[(carboxymethyl)(methyl)amino]methylidene}amino)phosphonate
O.O.O.O.[Na+].[Na+].CN(CC(O)=O)C(N)=NP([O-])([O-])=O
71519-72-7
C4H16N3Na2O9P
327.14
MFCD00150192
HUWYWJSJJDCZRQ-UHFFFAOYSA-L
creatine phosphate sodium salt,Sodium creatine phosphate dibasic tetrahydrate
131887655
disodium tetrahydrate ({amino[(carboxymethyl)(methyl)amino]methylidene}amino)phosphonate
O.O.O.O.[Na+].[Na+].CN(CC(O)=O)C(N)=NP([O-])([O-])=O
Formula Weight 327.14
Physical Form Crystalline Powder
Percent Purity ≥98%
Packaging Plastic bottle
Quantity 5g
Chemical Name or Material Creatine Phosphate Disodium Salt Tetrahydrate

EINECSNumber : 213-074-6

TSCA : No

Recommended Storage : Keep cold