Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Creatine phosphate disodium salt tetrahydrate, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Creatine phosphate disodium salt acts as a substrate for creatine kinase determinations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Creatine phosphate disodium salt acts as a substrate for creatine kinase determinations.
Solubility
Soluble in water.0.1 g/mL.
Notes
Incompatible with Strong bases. strong acids. Protect from Direct sunlight.
Creatine phosphate disodium salt acts as a substrate for creatine kinase determinations.
Solubility
Soluble in water.0.1 g/mL.
Notes
Incompatible with Strong bases. strong acids. Protect from Direct sunlight.
Specifications
Assay Percent Range | ≥98% |
Quantity | 1 g |
Solubility Information | Soluble in water.0.1g/mL. |
Formula Weight | 327.14 (255.07 Anhydrous) |
Percent Purity | ≥98% |
Physical Form | Powder |
Chemical Name or Material | Creatine Phosphate Disodium Salt Tetrahydrate |
Safety and Handling
EINECSNumber | 213-074-6 |
TSCA | No |
Recommended Storage | Keep cold |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction