Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Creatinine, 98%, Spectrum™

Click to view available options
Quantity:
10 g
25 g
100 g
1 kg
12 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Poly Pail
60-27-5
60-27-5
Packaging Poly Pail
Quantity 1 kg
CAS Max % 1
Percent Purity 0.98
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.