Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific Creatinine Reagents
Determine the quantity of creatinine in human serum.
Description
Determine the quantity of creatinine in human serum in vitro with Thermo Scientific Creatinine Reagents.
Creatinine is a non-protein nitrogenous metabolite that is cleared from the body by the kidney and is used to assess renal function. Elevated levels are observed in acute nephritis, renal failure, urinary tract obstruction, shock, dehydration, congestive heart failure and diabetes. This reagent offers up to 18 months of stability from manufacture date, and applications are available for most automated chemistry analyzers.Specifications
CE Marked | |
Serum | |
Reagent | |
Infinity Creatinine (Modified Jaffe) | |
0 to 20mg/dL | |
No | |
Stable Liquid |
Liquid | |
2°C to 8°C | |
Yes | |
Creatinine | |
Rate | |
2 x 125 mL | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title