Learn More
CREB Mouse anti-Human, Alexa Fluor™ 594, Clone: MM0704-13B29, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP236785AF594
Catalog No. NP23678A94
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
CREB Monoclonal antibody specifically detects CREB in Human samples. It is validated for Western Blot,Immunocytochemistry,Immunofluorescence.
Specifications
Specifications
CREB
MM0704-13B29
Western Blot, Immunocytochemistry/Immunofluorescence
active transcription factor CREB, cAMP responsive element binding protein 1, cAMP-response element-binding protein-1, cAMP-responsive element-binding protein 1, CREB, CREB-1, cyclic AMP-responsive element-binding protein 1, MGC9284, transactivator protein
Mouse
IgG2
Store at 4°C in the dark.
Monoclonal
This antibody recognizes human phosphorylated CREB S133 in WB.
Immunocytochemistry, Immunofluorescence, Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
CREB1
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with Phosphopeptide containing S133 site of human CREB).
0.1 mL
Primary
1385
Human