Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CREB (p Ser133) Mouse anti-Human, Biotin, Clone: MM0704-13B29, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236785B

Catalog No. NBP236785B


Add to cart

Description

Description

CREB p Ser133 Monoclonal antibody specifically detects CREB p Ser133 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

CREB (p Ser133)
MM0704-13B29
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
CREB1
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with Phosphopeptide containing S133 site of human CREB).
Protein G purified
Cancer, Chromatin Research, HIF Target Genes, Hypoxia, Immunology, Innate Immunity, Phospho Specific, Transcription Factors and Regulators
Primary
1385
Human
Western Blot, Immunocytochemistry, Immunofluorescence
Biotin
PBS with 0.05% Sodium Azide
active transcription factor CREB, cAMP responsive element binding protein 1, cAMP-response element-binding protein-1, cAMP-responsive element-binding protein 1, CREB, CREB-1, cyclic AMP-responsive element-binding protein 1, MGC9284, transactivator protein
Mouse
IgG2
0.1 ml
Store at 4C in the dark.
Monoclonal
This antibody recognizes human phosphorylated CREB S133 in WB.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit