Learn More
CREB (p Ser133) Mouse anti-Human, DyLight 350, Clone: MM0704-13B29, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236785UV
Catalog No. NBP236785UV
Add to cart
Description
Description
CREB p Ser133 Monoclonal antibody specifically detects CREB p Ser133 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
CREB (p Ser133)
MM0704-13B29
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
CREB1
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with Phosphopeptide containing S133 site of human CREB).
Protein G purified
Cancer, Chromatin Research, HIF Target Genes, Hypoxia, Immunology, Innate Immunity, Phospho Specific, Transcription Factors and Regulators
Primary
1385
Human
Western Blot, Immunocytochemistry, Immunofluorescence
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
active transcription factor CREB, cAMP responsive element binding protein 1, cAMP-response element-binding protein-1, cAMP-responsive element-binding protein 1, CREB, CREB-1, cyclic AMP-responsive element-binding protein 1, MGC9284, transactivator protein
Mouse
IgG2
0.1 ml
Store at 4C in the dark.
Monoclonal
This antibody recognizes human phosphorylated CREB S133 in WB.