Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sino Biological™ CREB1 (CREB) Recombinant Human Protein Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding human CREB1 isoform B (Met 1-Asp 327) fused to N-terminal polyhistidine-tagged MBP tag at N-terminus

Manufacturer:  Sino Biological™ 10074H10E5

 View more versions of this product

Catalog No. 10-074-H10E5


  Check Availability
Add to cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Expressed from DNA sequence encoding human CREB1 isoform B (Met 1-Asp 327) fused to N-terminal polyhistidine-tagged MBP tag at N-terminus

  • 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween80
Description & Specifications

Specifications

Lyophilized
78.7kd
5μg
−20°C
CAMP responsive element binding protein 1
Assay Standard
E.coli
N-His.MBP Tag
1385
≥90% by SDS-PAGE
Human
Recombinant
CREB1
Transcription Factors
CREB1 (CREB) Recombinant Human Protein
Provide Content Correction

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit