Sino Biological™ CREB1 (CREB) Recombinant Human Protein Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding human CREB1 isoform B (Met 1-Asp 327) fused to N-terminal polyhistidine-tagged MBP tag at N-terminus

Manufacturer: Sino Biological™ 10074H10E25

 View more versions of this product

Catalog No. 10-074-H10E25

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Quantity 5 x 5μg
    Source Human
    Storage Requirements −20°C
    Common Name CAMP responsive element binding protein 1
    Format Lyophilized
    Molecular Weight 78.7kd
    Purity ≥90% by SDS-PAGE
    Structural Form Recombinant
    Gene Symbol CREB1
    Protein Tag N-His.MBP Tag
    Validated Application Assay Standard
    Protein Family Transcription Factors
    Expression System E.coli
    Product Type CREB1 (CREB) Recombinant Human Protein
    Gene ID (Entrez) 1385

    Expressed from DNA sequence encoding human CREB1 isoform B (Met 1-Asp 327) fused to N-terminal polyhistidine-tagged MBP tag at N-terminus

    • 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween80

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit