Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CREB1, Rabbit anti-Human, FITC, Clone: 4D11, Abnova™

Rabbit Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB18875

Catalog No. 89-335-709


Add to cart

Specifications

Specifications

CREB1
4D11
Flow Cytometry (5 uL/106 cells) The optimal working dilution should be determined by the end user.
cAMP responsive element binding protein 1
CREB|MGC9284
Rabbit
IgG1, kappa
RUO
Antibody
Monoclonal
Human, Mouse
Flow Cytometry
FITC
In PBS, pH 7.4 (0.2% BSA, 0.09% sodium azide).
Liquid
CREB1
A synthetic phosphopeptide corresponding to residues surrounding S133 of human CREB1.
100 Reactions
Store at 4°C.
Primary
1385
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit