Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Human CREB3L2 His Protein
Highly purified. Generating reliable and reproducible results.
Supplier: Novus Biologicals™ NBP2519270.1MG
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-378 of Human CREB3L2 The Recombinant Human CREB3L2 Protein is derived from E. coli. The Recombinant Human CREB3L2 Protein has been validated for the following applications: SDS-Page.Specifications
0.25mg/mL | |
PBS (pH 7.4) containing 30% glycerol, 1mM DTT. | |
TMW: 44kDa | |
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. | |
CREB3L2 | |
Recombinant |
SDS-PAGE | |
64764 | |
0.1mg | |
basic transcription factor 2, BBF2 human homolog on chromosome 7, BBF2H7TCAG_1951439, B-ZIB transcription factor, cAMP responsive element binding protein 3-like 2, cAMP-responsive element-binding protein 3-like protein 2, cyclic AMP-responsive element-binding protein 3-like protein 2, FUS/BBF2H7 protein, MGC131709, MGC71006 | |
Unconjugated | |
>85%, by SDS-PAGE |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For research use only and is not approved for use in humans or in clinical diagnosis
Spot an opportunity for improvement?
Provide Content Correction