Learn More
Aviva Systems Biology Corp CREBZF Antibody - middle region (ARP47526_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP47526_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CREBZF. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CREBZF. Alternate names: ZF, SMILEApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59577