Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CREBZF Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 190701AP

Catalog No. 50-173-2333


Add to cart

Description

Description

ZHANGFEI, also known as ZF, is a basic region-leucine zipper transcriptional factor that has been implicated in herpes virus infection cycle and related cellular processes. ZHANGFEI protein has a negatively charged active N-terminus, a leucine zipper of six heptad leucine repeats separated by a conserved six amino acid spacer, a basic domain and a bZIP region. ZHANGFEI strongly activates transcription and suppresses the expression of HSV proteins in cells infected with the virus in a HCFC1 dependent manner. It also suppresses the HCFC1 dependent transcriptional activation by CREB3 and reduces the amount of CREB3 in the cell.
Specifications

Specifications

CREBZF
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q91ZR3, Q9NS37
CREBZF, SMILE, ZF, Zhangfei
Rabbit
IgG
150 μL
-20°C
Primary
233490, 293112, 58487
Western Blot
Unconjugated
CREBZF
Liquid
CREBZF
CREBZF Fusion Protein Ag13625
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit