Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CREG Mouse anti-Human, Alexa Fluor™ 532, Clone: 1C6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP242689AF532

Catalog No. NP242689A32


Add to cart

Description

Description

CREG Monoclonal antibody specifically detects CREG in Human samples. It is validated for Western Blot,ELISA.
Specifications

Specifications

CREG
1C6
Western Blot, ELISA
cellular repressor of E1A-stimulated genes, cellular repressor of E1A-stimulated genes 1CREG, protein CREG1
Mouse
IgG2b Kappa
Store at 4°C in the dark.
Monoclonal
Human
ELISA, Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
CREG1
Recombinant human CREG1 (32-220aa) purified from E. coli
0.1 mL
Primary
8804
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit