Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CREG Antibody (1C6), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242689AF750
Description
CREG Monoclonal specifically detects CREG in Human samples. It is validated for Western Blot, ELISA.Specifications
CREG | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
CREG1 | |
Recombinant human CREG1 (32-220aa) purified from E. coli | |
Cancer, Cell Cycle and Replication | |
8804 | |
Store at 4°C in the dark. |
Western Blot, ELISA | |
1C6 | |
Western Blot, ELISA | |
cellular repressor of E1A-stimulated genes, cellular repressor of E1A-stimulated genes 1CREG, protein CREG1 | |
Mouse | |
100 μL | |
Primary | |
Human | |
IgG2b κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction