Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRHBP Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF2796
This item is not returnable.
View return policy
Description
CRHBP Polyclonal specifically detects CRHBP in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
CRHBP | |
Polyclonal | |
Unconjugated | |
P24387.2 | |
CRHBP | |
Mouse myeloma cell line NS0-derived recombinant human CRHBP Tyr25-Leu322 Accession # P24387.2 | |
100 μg | |
Primary | |
Detects human CRHBP in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant mouse CRHBP is observed. | |
Human | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
corticotropin releasing hormone binding protein, corticotropin-releasing factor-binding protein, Corticotropin-releasing hormone-binding protein, CRFBP, CRFBPcorticotropin releasing hormone-binding protein, CRF-BPCRF-binding protein, CRH-BP | |
Goat | |
Affinity Purified | |
RUO | |
1393 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only