Learn More
Sklar InstrumentsSupplier Diversity Partner Crile Needle Holder, Sklar, 6 in., Blunt, Straight, Serrated, Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 201160
Crile Needle Holder, Sklar, 6 in., Blunt, Straight, Serrated, Stainless Steel. Sklar's Crile Needle Holder is used to hold and guide intermediate weight needles during suturing. This product is straight with serrated tips, and an overall length of 6 inches.
Catalog No. 50-143-7444