Learn More
Sklar InstrumentsSupplier Diversity Partner Crile-Wood Needle Holder, Sklar, 8 in., Blunt, Straight, Cross-Serrated with Groove, Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 201280
Crile-Wood Needle Holder, Sklar, 8 in., Blunt, Straight, Cross-Serrated with Groove, Stainless Steel. Sklar's Crile-Wood Needle Holder is used to hold and guide small to medium size needles and suture materials. The Crile-Wood Needle Holder has a gently tapered, blunt tip and is generally more delicate than the Mayo-Hegar but slightly less delicate than Webster, Halsey, or DeBakey needle holders. The ratcheted finger ring handle aids in securing needles for both continuous and interrupted closures. This product is straight with cross-serrated tips, center notch and an overall length of 8 inches.
Catalog No. 50-143-7448