Learn More
CRIM1 Mouse anti-Human, Alexa Fluor™ 594, Clone: MM0202-16G1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212207AF594
Catalog No. NP212207A94
Add to cart
Description
Description
CRIM1 Monoclonal antibody specifically detects CRIM1 in Human samples. It is validated for Western Blot,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
CRIM1
MM0202-16G1
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
CRIM-1, cysteine rich transmembrane BMP regulator 1 (chordin-like), cysteine-rich motor neuron 1, cysteine-rich motor neuron 1 protein, Cysteine-rich repeat-containing protein S52, MGC138194, S52
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
CRIM1
Human CRIM1
0.1 mL
Primary
51232