Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRIM1 Mouse anti-Human, Clone: MM0202-16G1, Novus Biologicals™
Mouse Monoclonal Antibody
$177.16 - $493.00
Specifications
Antigen | CRIM1 |
---|---|
Clone | MM0202-16G1 |
Concentration | LYOPH |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
CRIM1 Monoclonal specifically detects CRIM1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CRIM1 | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
Q9NZV1 | |
51232 | |
IgG1 | |
Protein G purified |
MM0202-16G1 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
CRIM-1, cysteine rich transmembrane BMP regulator 1 (chordin-like), cysteine-rich motor neuron 1, cysteine-rich motor neuron 1 protein, Cysteine-rich repeat-containing protein S52, MGC138194, S52 | |
CRIM1 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title