Learn More
Aviva Systems Biology Corp CRIP1 Antibody - N-terminal region (ARP51614_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP51614_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CRIP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CRIP1. Alternate names: CRHP, CRIP, CRP1, CRP-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-60569