Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRIP2 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259094

Catalog No. NBP259094


Add to cart

Description

Description

CRIP2 Polyclonal antibody specifically detects CRIP2 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

CRIP2
Unconjugated
Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide
CRIP, CRP-2, CRP2ESP1, cysteine-rich protein 2, Cystein-rich intestinal protein, Protein ESP1
Rabbit
IgG
100 ul
Store at 4C.
Polyclonal
Human, Mouse
Western Blot, Immunoprecipitation
Western Blot, Immunoprecipitation
Affinity Purified
CRIP2
Maps to a region between 158 and 208
Immunogen affinity purified
RUO
Primary
1397
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only