Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRIPAK Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549510

Catalog No. PA549510


Add to cart

Description

Description

Negative regulator of PAK1. It has been suggested that the lost of CRIPAK in breast tumors might contribute to hormonal independence.
Specifications

Specifications

CRIPAK
Polyclonal
Unconjugated
CRIPAK
Liquid
CRIPAK
KLH conjugated synthetic peptide between 134-165 amino acids from the Central region of human CRIPAK.
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q8N1N5
Cysteine-rich PAK1 inhibitor, CRIPak, CRIPAK
Rabbit
IgG
200 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
285464
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit