Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRIPT Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA528958

Catalog No. PIPA528958


Only null left
Add to Cart

Description

Description

Recommended positive controls: mouse brain. Predicted reactivity: Mouse (99%), Rat (99%), Zebrafish (93%), Xenopus laevis (93%), Chicken (96%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Involved in the cytoskeletal anchoring of DLG4 in excitatory synapses.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CRIPT
Polyclonal
Unconjugated
CRIPT
1200020A08Rik; AA589469; CRIPT; CXXC repeat containing interactor of PDZ3 domain; cysteine-rich interactor of PDZ three; Cysteine-rich interactor of PDZ3; cysteine-rich PDZ-binding protein; HSPC139; postsynaptic protein CRIPT; SSMDF
Rabbit
Antigen affinity chromatography
RUO
56724, 9419
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
O70333, Q9P021
CRIPT
Full length human CRIPT Recombinant protein.
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.