Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CRISP-1 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $468.00
Specifications
Antigen | CRISP-1 |
---|---|
Clone | 840017 |
Dilution | Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Neutralization 10-20 ug/mL |
Applications | Western Blot, Immunohistochemistry, Neutralization |
Classification | Monoclonal |
Description
CRISP-1 Monoclonal specifically detects CRISP-1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.Specifications
CRISP-1 | |
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Neutralization 10-20 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Acidic epididymal glycoprotein homolog, acidic epididymal glycoprotein-like 1, AEGL1, AEG-related protein, ARPAEG-like protein, CRISP1, CRISP-1AEGL1HSCRISP1D, cysteine-rich secretory protein 1, cysteine-rich secretory protein-1 delta, HSCRISP1G, HUMARP | |
Crisp1 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
840017 | |
Western Blot, Immunohistochemistry, Neutralization | |
Unconjugated | |
Rat | |
Mouse | |
Q03401 | |
167 | |
Mouse myeloma cell line NS0-derived recombinant mouse CRISP-1 Gln20-His244 Accession # Q03401 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse CRISP-1 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title