Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CRISP3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA545558
Description
Peptide sequence: PARNMLKMEW NKEAAANAQK WANQCNYRHS NPKDRMTSLK CGENLYMSSA Sequence homology: Human: 100%; Yeast: 77%.
Cysteine-rich secretory protein 3 (CRISP-3; also known as SGP28) is a glycoprotein that belongs to the family of cysteine-rich secretory proteins (CRISPs) which was originally discovered in human neutrophilic granulocytes. CRISP-3 is also widely distibuted in exocrine glands (salivary glands, pancreas and prostate), eosinophilic granulocytes and to a lower level in epididymis, ovary, thymus and colon. The presence of CRISP-3 in neutrophils, eosinophils and in exocrine secretions indicates a role in innate host defense. The antibody has been raised against recombinant C-terminally truncated form of CRISP-3 and recognizes both the N-glycosylated and non-glycosylated form of the mature protein.
Specifications
CRISP3 | |
Polyclonal | |
Unconjugated | |
Crisp3 | |
32 kDa epididymal protein; Acidic epididymal glycoprotein; Acidic epididymal glycoprotein 2; acidic epididymal glycoprotein D/E; Aeg; Aeg2; Aeg-2; Crisp1; Crisp-1; Crisp3; CRISP-3; CRISP-3MGC126588; CRS3; cysteine rich secretory protein 3; cysteine-rich secretory protein 1; Cysteine-rich secretory protein 3; cysteine-rich secretory protein-3; dJ442L6.3; epididymal glycoprotein; protein D; protein E; protein IV; SCP; SCP 2; SGP28; Sialoprotein; specific granule protein (28 kDa); specific granule protein of 28 kDa; Sperm-coating glycoprotein; sperm-coating glycoprotein 2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10321 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P54108 | |
Crisp3 | |
Synthetic peptide directed towards the N-terminal of human CRISP3 (aa 56-105). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction