Learn More
CRISP3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 148471AP
Catalog No. 50-172-9774
Add to cart
Description
Description
This antibody can recognize both CRISP2 and CRISP3.
The function of this protein remains unknown.
Specifications
Specifications
CRISP3
0.42 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P54108, Q03402
Aeg2, CRISP 3, CRISP3, CRS3, SGP28
Rabbit
IgG
150 μL
-20°C
Primary
10321, 11572
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
CRISP3
Liquid
CRISP3
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse