Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CRISPLD2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543477
Description
Peptide sequence: MSCVLGGVIP LGLLFLVCGS QGYLLPNVTL LEELLSKYQH NESHSRVRRA Sequence homology: Human: 100%; Mouse: 86%; Rat: 86%.
CRISPLD2 promotes matrix assembly.
Specifications
CRISPLD2 | |
Polyclonal | |
Unconjugated | |
CRISPLD2 | |
1810049K24Rik; coffeecrisp; CRISP11; CRISP-11; CRISPLD2; cysteine rich secretory protein LCCL domain containing 2; cysteine-rich secretory protein 11; cysteine-rich secretory protein LCCL domain containing 2; cysteine-rich secretory protein LCCL domain-containing 2; late gestation lung protein 1; LCCL domain containing cysteine-rich secretory protein 2; LCCL domain-containing cysteine-rich secretory protein 2; Lcrisp2; Lgl1; testis secretory sperm-binding protein Li 207a; trypsin inhibitor; UNQ2914/PRO1156/PRO9783 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
83716 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9H0B8 | |
CRISPLD2 | |
Synthetic peptide directed towards the N-terminal of human CRISPLD2 (aa 1-50). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction