Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRISPR-Cas9 Mouse anti-Bacteria, Alexa Fluor™ 750, Clone: 6G12, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP252398AF750

Catalog No. NP22398A70


Add to cart

Description

Description

CRISPR-Cas9 Monoclonal antibody specifically detects CRISPR-Cas9 in Bacteria samples. It is validated for Western Blot,Immunocytochemistry,Immunofluorescence.
Specifications

Specifications

CRISPR-Cas9
6G12
Western Blot, Immunocytochemistry/Immunofluorescence
Cas9, CRISPR, CRISPR-associated protein 9 nuclease
Recombinant C-terminal fragment of S.pyogenes CRISPR/Cas9. [UniProtNo. Q99ZW2]
0.1 mL
Store at 4°C in the dark.
Monoclonal
Cas9 from Streptococcus pyogene.
Immunocytochemistry, Immunofluorescence, Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
Mouse
IgG1 Kappa
Gene Editing, Gene Knockouts, Gene Therapy
Primary
901176
Bacteria
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit