Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRISPR-Cas9 Rabbit anti-Bacteria, Alexa Fluor 700, Polyclonal,Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP266379AF700

Catalog No. NBP379AF700


Add to cart

Description

Description

CRISPR-Cas9 Polyclonal antibody specifically detects CRISPR-Cas9 in Bacteria samples. It is validated for Western Blot.
Specifications

Specifications

CRISPR-Cas9
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
Cas9, CRISPR, CRISPR-associated protein 9 nuclease
Full length S. pyogenes Cas9 protein [UniProt Q99ZW2]
Immunogen affinity purified
Gene Editing, Gene Knockouts, Gene Therapy
Primary
Cas9 protein from Streptococcus pyogene serotype M1.
Western Blot
Western Blot
Affinity Purified
Rabbit
IgG
0.1mL
Store at 4°C in the dark.
Polyclonal
Bacteria
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit