Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRISPR-Cas9 Rabbit anti-Bacteria, DyLight 405, Polyclonal,Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP266379V
Description
CRISPR-Cas9 Polyclonal antibody specifically detects CRISPR-Cas9 in Bacteria samples. It is validated for Western Blot.Specifications
CRISPR-Cas9 | |
DyLight 405 | |
Affinity Purified | |
Cas9, CRISPR, CRISPR-associated protein 9 nuclease | |
Full length S. pyogenes Cas9 protein [UniProt Q99ZW2] | |
Polyclonal | |
Immunogen affinity purified | |
Gene Editing, Gene Knockouts, Gene Therapy | |
Bacteria |
Western Blot | |
Western Blot | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Rabbit | |
IgG | |
Primary | |
0.1mL | |
Store at 4°C in the dark. | |
Cas9 protein from Streptococcus pyogene serotype M1. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title