Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRISPR-Cpf1, Mouse anti-Bacteria, Alexa Fluor 350, Clone: 2D5-6G11, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP278819AF350
Description
CRISPR-Cpf1 Monoclonal antibody specifically detects CRISPR-Cpf1 in Bacteria samples. It is validated for Western Blot.Specifications
CRISPR-Cpf1 | |
2D5-6G11 | |
Western Blot | |
Conjugated, Purified | |
Mouse | |
IgG1 Kappa | |
0.1 mL | |
Store at 4°C in the dark. | |
Monoclonal | |
Bacteria |
Western Blot | |
Alexa Fluor 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Cas12a, Clustered regularly interspaced short palindromic repeat (CRISPR) associated Cas12a, Cpf1, CRISPR-associated endonuclease Cas12a, CRISPR-associated endonuclease Cpf1, CRISPR-Cas12a, FnCas12a, FnCpf1 | |
Partial recombinant protein made to an N-terminal portion of the Lachnospiraceae bacterium CRISPR-Cpf1 protein | |
Protein A or G purified | |
RUO | |
Primary | |
This antibody is specific for Lachnospiraceae bacterium CRISPR-Cpf1. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title