Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRISPR-Cpf1 Rabbit anti-Bacteria, Alexa Fluor™ 647, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP262735AF647

Catalog No. N262735A647


Add to cart

Description

Description

CRISPR-Cpf1 Polyclonal antibody specifically detects CRISPR-Cpf1 in Bacteria samples. It is validated for Western Blot.
Specifications

Specifications

CRISPR-Cpf1
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
Cas12a, Clustered regularly interspaced short palindromic repeat (CRISPR) associated Cas12a, Cpf1, CRISPR-associated endonuclease Cas12a, CRISPR-associated endonuclease Cpf1, CRISPR-Cas12a, FnCas12a, FnCpf1
Partial recombinant Francisella tularensis protein (between amino acids 600-850) [NCBI WP_003034647.1]
Immunogen affinity purified
Store at 4°C in the dark.
Polyclonal
Western Blot
Western Blot
Affinity Purified, Conjugated
Rabbit
IgG
0.1 mL
Primary
Bacteria
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit