Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRISPR-Cpf1 Rabbit anti-Bacteria, DyLight™ 550, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP262735R
Description
CRISPR-Cpf1 Polyclonal antibody specifically detects CRISPR-Cpf1 in Bacteria samples. It is validated for Western Blot.Specifications
CRISPR-Cpf1 | |
DyLight 550 | |
Affinity Purified,Conjugated | |
Cas12a, Clustered regularly interspaced short palindromic repeat (CRISPR) associated Cas12a, Cpf1, CRISPR-associated endonuclease Cas12a, CRISPR-associated endonuclease Cpf1, CRISPR-Cas12a, FnCas12a, FnCpf1 | |
Partial recombinant Francisella tularensis protein (between amino acids 600-850) [NCBI WP_003034647.1] | |
Polyclonal | |
Immunogen affinity purified | |
Store at 4°C in the dark. |
Western Blot | |
Western Blot | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Rabbit | |
IgG | |
Primary | |
0.1 mL | |
Bacteria |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title