Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Crk Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP257342

Catalog No. NBP257342


Add to cart

Description

Description

Crk Polyclonal antibody specifically detects Crk in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Crk
Unconjugated
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
adapter molecule crk, avian sarcoma virus CT10 (v-crk) oncogene homolog, CRKII, p38, Proto-oncogene c-Crk, v-crk avian sarcoma virus CT10 oncogene homolog, v-crk sarcoma virus CT10 oncogene homolog (avian)
Rabbit
IgG
100 ul
Apoptosis, Ovarian Carcinoma Cell Markers
Primary
1398
Human
Immunocytochemistry, Immunofluorescence
Immunocytochemistry/Immunofluorescence 1-4 ug/ml
Affinity Purified
CRK
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:DTTTLIEPVSRSRQGSGVILRQEEAEYVRALFDFNGNDEEDLPF
Affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only