Learn More
CrkL Mouse anti-Human, Alexa Fluor™ 350, Clone: MM0707-9F39, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP236786AF350
Catalog No. NP236786A30
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
CrkL Monoclonal antibody specifically detects CrkL in Human samples. It is validated for Western Blot.
Specifications
Specifications
CrkL
MM0707-9F39
Western Blot
crk-like protein, v-crk avian sarcoma virus CT10 oncogene homolog-like, v-crk sarcoma virus CT10 oncogene homolog (avian)-like
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
This antibody recognizes human CrkL with phospho-Y207 in direct ELISAs and Western blots.
Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
CRKL
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with Phosphopeptide containing human CrkL Y207).
0.1 mL
Primary
1399
Human