Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CrkL (p Tyr207) Mouse anti-Human, Biotin, Clone: MM0707-9F39, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236786B

Catalog No. NBP236786B


Add to cart

Description

Description

CrkL p Tyr207 Monoclonal antibody specifically detects CrkL p Tyr207 in Human samples. It is validated for Western Blot.
Specifications

Specifications

CrkL (p Tyr207)
MM0707-9F39
Western Blot
Purified
CRKL
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with Phosphopeptide containing human CrkL Y207).
Protein G purified
Cancer, Phospho Specific, Signal Transduction
Primary
1399
Human
Western Blot
Biotin
PBS with 0.05% Sodium Azide
crk-like protein, v-crk avian sarcoma virus CT10 oncogene homolog-like, v-crk sarcoma virus CT10 oncogene homolog (avian)-like
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
This antibody recognizes human CrkL with phospho-Y207 in direct ELISAs and Western blots.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit