Learn More
CrkL (p Tyr207) Mouse anti-Human, DyLight 550, Clone: MM0707-9F39, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236786R
Catalog No. NBP236786R
Add to cart
Description
Description
CrkL p Tyr207 Monoclonal antibody specifically detects CrkL p Tyr207 in Human samples. It is validated for Western Blot.
Specifications
Specifications
CrkL (p Tyr207)
MM0707-9F39
Western Blot
Purified
CRKL
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with Phosphopeptide containing human CrkL Y207).
Protein G purified
Cancer, Phospho Specific, Signal Transduction
Primary
1399
Human
Western Blot
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
crk-like protein, v-crk avian sarcoma virus CT10 oncogene homolog-like, v-crk sarcoma virus CT10 oncogene homolog (avian)-like
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
This antibody recognizes human CrkL with phospho-Y207 in direct ELISAs and Western blots.