Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRLF3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548873

Catalog No. PA548873

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with mouse based on sequence homology.

May play a role in the negative regulation of cell cycle progression.
Specifications

Specifications

CRLF3
Polyclonal
PBS with 0.09% sodium azide
Q8IUI8
Cytokine receptor-like factor 3, Cytokine receptor-like molecule 9, CREME-9, Cytokine receptor-related protein 4, Type I cytokine receptor-like factor p48, CRLF3, CREME9, CRLM9, CYTOR4, P48
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51379
Western Blot
Unconjugated
CRLF3
Liquid
CRLF3
KLH conjugated synthetic peptide between 305-331 amino acids from the C-terminal region of human CRLF3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit