Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CRNKL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570817
Description
This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 86%; Rat: 100%.
CRNKL1 is a protein coding gene.
Specifications
CRNKL1 | |
Polyclonal | |
Unconjugated | |
CRNKL1 | |
1200013P10Rik; 5730590A01Rik; C80326; CGI-201; CLF; Clf1; Crn; Crn, crooked neck-like 1; Crn, crooked neck-like 1 (Drosophila); Crnkl1; Crooked neck homolog; crooked neck pre-mRNA splicing factor 1; crooked neck pre-mRNA splicing factor-like 1; Crooked neck protein; crooked neck-like 1; crooked neck-like 1 protein; crooked neck-like protein 1; crooked neck-like protein 1-like; crooked neck-like protein 1-like protein; hCrn; MSTP021; SYF3; SYF3 pre-mRNA-splicing factor | |
Rabbit | |
Affinity chromatography | |
RUO | |
51340 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9BZJ0 | |
CRNKL1 | |
synthetic peptide directed towards the C-terminal region of human CRNKL1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction