Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CROCCL2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP17947020UL

 View more versions of this product

Catalog No. NBP17947020


Add to cart

Description

Description

CROCCL2 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

CROCCL2
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
ciliary rootlet coiled-coil, rootletin pseudogene 3, CROCCL2, dJ798A10.2, KIAA0445, KIAA1922
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
114819
Western Blot
Western Blot 1:1000
Affinity Purified
CROCCP3
Synthetic peptide directed towards the N terminal of human CROCCL2The immunogen for this antibody is CROCCL2. Peptide sequence NPEKDQVNTDLTEKLEALGTWHSPSCRTQSGVQLSGSERTADDSFGSLRG.
47 kDa
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit