Alfa Aesar™ Croconic acid, 98%

Manufacturer: Alfa Aesar™ B2180906

Quantity 5g

 View more versions of this product

Catalog No. AAB2180906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 488-86-8
    Molecular Formula C5H2O5
    Formula Weight 142.07
    MDL Number MFCD00181389
    Synonym 4,5-Dihydroxy-4-cyclopentene-1,2,3-trione; 1,2-Dihydroxy-3,4,5-trioxocyclopent-1-ene
    Chemical Name or Material Croconic acid

    Specifications

    Formula Weight 142.07
    Odor Odorless
    MDL Number MFCD00181389
    Quantity 5g
    Molecular Formula C5H2O5
    CAS 488-86-8
    Synonym 4,5-Dihydroxy-4-cyclopentene-1,2,3-trione; 1,2-Dihydroxy-3,4,5-trioxocyclopent-1-ene
    Health Hazard 1 Warning
    Melting Point ≈280°C (dec.)
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Chemical Name or Material Croconic acid
    Assay 98%

    Description

    Applications
    Croconic acid is used to prepare 1,3-bis-(2-dimethylamino-5-thienyl)croconine by reacting with dimethyl-thiophen-2-yl-amine. It is also used in organic synthesis and employed as a dye for biological research purposes. Further, it is involved in the preparation of ethers such as dimethyl croconate. In addition to this, it acts as a ligand and form coordination complexes with metals such as barium, copper, silver and lead.

    Solubility
    Soluble in water and ethanol.

    Notes
    Light sensitive. Incompatible with strong oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit