Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Croconic acid disodium salt, 97%

Click to view available options
Quantity:
0.25g
1g
14379-00-1
C5Na2O5
186.03
MFCD00191954
OQXLFPHHAAAVKQ-UHFFFAOYSA-L
croconic acid disodium salt,croconic acid, disodium salt,disodium trioxocyclopent-1-ene-1,2-bis olate,sodium 3,4,5-trioxocyclopent-1-ene-1,2-bis olate,4,5-dihydroxy-4-cyclopentene-1,2,3-trione disodium salt
12120285
disodium;3,4,5-trioxocyclopentene-1,2-diolate
C1(=C(C(=O)C(=O)C1=O)[O-])[O-].[Na+].[Na+]
14379-00-1
C5Na2O5
186.03
MFCD00191954
OQXLFPHHAAAVKQ-UHFFFAOYSA-L
croconic acid disodium salt,croconic acid, disodium salt,disodium trioxocyclopent-1-ene-1,2-bis olate,sodium 3,4,5-trioxocyclopent-1-ene-1,2-bis olate,4,5-dihydroxy-4-cyclopentene-1,2,3-trione disodium salt
12120285
disodium;3,4,5-trioxocyclopentene-1,2-diolate
C1(=C(C(=O)C(=O)C1=O)[O-])[O-].[Na+].[Na+]
Quantity 1g
Melting Point >300°C
Formula Weight 186.03
Percent Purity 97%
Chemical Name or Material Croconic acid disodium salt