Learn More
Cromakalim, Tocris
Kir6 (KATP) channel opener
Manufacturer:  R&D Systems 1377/10
Quantity 10mg
 View more versions of this product
Catalog No. 13-771-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>99%
286.33
Cromakalim
C16H18N2O3
trans-3, 4-dihydro-3-hydroxy-2, 2-dimethyl-4-(2-oxo-1-pyrrolidinyl)-2H-1-benzopyran-6-carbonitrile
Specifications
10mg
>99%
286.33
Cromakalim
94470-67-4
C16H18N2O3
trans-3, 4-dihydro-3-hydroxy-2, 2-dimethyl-4-(2-oxo-1-pyrrolidinyl)-2H-1-benzopyran-6-carbonitrile
Safety and Handling
Recommended Storage : Store at Room Temperature