Learn More
Croscarmellose Sodium, NF, Spectrum™
Click to view available options
Quantity:
500 g
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
74811-65-7
74811-65-7
C1366, 74811-65-7
Spectrum™ Croscarmellose Sodium, NF is used commonly as a FDA-approved disintegrant in pharmaceutical manufacturing. The NF grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Poly Bottle
Loss on Drying 0.1
Ignition Residue 14.0-28%
Quantity 500 g
CAS Max % 1